Learn More
TARGETMOL CHEMICALS INC Angiotensin III human mouse
SDP
Catalog No. 502876294
Encompass_Preferred
Also available in 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Angiotensin III, human, mouse (Angiotensin III, human, mouse(3TFA))(3TFA) is an endogenous angiotensin II receptor (AT2R) agonist(IC50s of 0.648 nM). Purity 100%
Catalog No. 50-287-6294 Supplier TARGETMOL CHEMICALS INC Supplier No. T757750MG
Also available in 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Angiotensin III, human, mouse (Angiotensin III, human, mouse(3TFA))(3TFA) is an endogenous angiotensin II receptor (AT2R) agonist(IC50s of 0.648 nM). Purity 100%