Learn More
Medchemexpress LLC HY-113035A 10mg Medchemexpress, Angiotensin III (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY113035A10MG
Encompass_Preferred
Medchemexpress, HY-113035A 10mg Angiotensin III (TFA) CAS: Angiotensin III (TFA) is an angiotensin 1 (AT1) and AT2 receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9219