Learn More
CPC Scientific H-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Leu-Val-Tyr-Ser-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific RENI004A
Encompass
Renin substrate; lyophilized solid. It is split by renin to generate angiotensin I and is readily cleaved by human cathepsin D.ONE-LETTER SEQUENCE: DRVYIHPFHLLVYSMOLECULAR FORMULA: C85H123N21O20MOLECULAR WEIGHT:1759.01STORAGE CONDITIONS: -20 5CSYNONYMS: Angiotensinogen 1-14 Renin Substrate porcineRESEARCH AREA: CardiovascularREFERENCES: R. Kageyama et al., Biochemistry, 23, 3603 (1984)
Catalog No. 50-211-0360