Learn More
Surgical Design™ Angle Finder Ruler
Catalog No. 21124920 Shop All Surgical Design Products
Catalog No. 21-124-920 Supplier Surgical Design™ Supplier No. MT004
Add to Cart
Add to Cart
Angle finder ruler measures, transfers, and replicates angles quickly for technical, laboratory, construction, and workshop applications.
  • Dual-scale protractor design for accurate angle measurement and marking
  • Adjustable swinging arm easily locks in place for repeatable angle transfer
  • Clear, easy-read graduations for fast and precise alignment
  • Lightweight and durable construction suitable for field or bench use
  • Ideal for drafting, machining, woodworking, QA inspections, and scientific measurements
  • Compact form factor fits conveniently into toolkits, lab drawers, and field bags