Learn More
Grainger Angle Grinder 10.5 A Rat Tail 5.9 lb
Catalog No. 502753406
Encompass_Preferred
The Makita 10.5 Amp 4-1/2 in. Angle Grinder is a powerful electric angle grinder designed for cutting, grinding and sanding applications. It features a 10.5 Amp motor that delivers up to 11,000 RPM for fast material removal. The compact and ergonomic design reduces fatigue during extended use. It has a tool-less wheel guard for quick adjustments and a side handle that can be installed on either side. Additional features include a lock-on slide switch, AC/DC switch for use with generators, and a protective zigzag varnish sealed windings to resist dust and debris. This angle grinder is ideal for metal fabrication, concrete work, tile, auto body work and more.
Catalog No. 50-275-3406 Supplier Grainger Supplier No. 14F099
The Makita 10.5 Amp 4-1/2 in. Angle Grinder is a powerful electric angle grinder designed for cutting, grinding and sanding applications. It features a 10.5 Amp motor that delivers up to 11,000 RPM for fast material removal. The compact and ergonomic design reduces fatigue during extended use. It has a tool-less wheel guard for quick adjustments and a side handle that can be installed on either side. Additional features include a lock-on slide switch, AC/DC switch for use with generators, and a protective zigzag varnish sealed windings to resist dust and debris. This angle grinder is ideal for metal fabrication, concrete work, tile, auto body work and more.