Learn More
Grainger Angle Grinder 4-1/2 7 A 10 000 RPM 120V
Catalog No. 502751748
Encompass_Preferred
The Makita 7.5 Amp 4-1/2 in. Angle Grinder is a powerful, compact grinder ideal for metalworking, cutting, grinding and sanding applications. It features a 7.5 Amp motor that delivers 11,000 RPM for fast material removal. The small diameter barrel grip provides ease of handling. It has a lock-on slide switch for continuous operation. The grinder has a 4-1/2 in. wheel capacity and is double insulated for user protection. It weighs just 5 lbs for reduced fatigue during extended use. This angle grinder is backed by a 1 year warranty from Makita.
Catalog No. 50-275-1748 Supplier Grainger Supplier No. 14F047
The Makita 7.5 Amp 4-1/2 in. Angle Grinder is a powerful, compact grinder ideal for metalworking, cutting, grinding and sanding applications. It features a 7.5 Amp motor that delivers 11,000 RPM for fast material removal. The small diameter barrel grip provides ease of handling. It has a lock-on slide switch for continuous operation. The grinder has a 4-1/2 in. wheel capacity and is double insulated for user protection. It weighs just 5 lbs for reduced fatigue during extended use. This angle grinder is backed by a 1 year warranty from Makita.