Learn More
Grainger Angle Grinder 5 in No Load RPM 11500
Supplier:  Grainger 14F050
Encompass_Preferred
The Makita 4-1/2" Angle Grinder, model XAG03Z, is a 13 amp angle grinder with a no-load speed of 10,000 RPM. It has a small diameter barrel grip for ease of handling and comfort. The motor is engineered for improved performance and overload protection. It has an AC/DC switch for use with generators. The grinding wheel guard can be installed tool-free for quick adjustments. Overall, this Makita angle grinder is a powerful, durable tool designed for metal grinding, cutting and sanding applications.
Catalog No. 50-275-1771