Learn More
Grainger Angle Grinder 5 in No Load RPM 11000
Catalog No. 502749889
Encompass_Preferred
The Makita 4-1/2" Angle Grinder is compact and lightweight, ideal for cutting, grinding, and sanding. It features a 5.5 amp motor delivering 11,000 RPM for fast material removal. The small barrel grip design ensures easy handling. It has an AC/DC switch for alternative power sources and a lock-on slide switch for continuous operation. The spindle lock allows quick wheel changes. It includes a side handle, wheel guard, and wrench, and weighs just 3.5 lbs, reducing fatigue during extended use. This versatile grinder is suitable for metalworking, concrete grinding, and more.
Catalog No. 50-274-9889 Supplier Grainger Supplier No. 14F098
The Makita 4-1/2" Angle Grinder is compact and lightweight, ideal for cutting, grinding, and sanding. It features a 5.5 amp motor delivering 11,000 RPM for fast material removal. The small barrel grip design ensures easy handling. It has an AC/DC switch for alternative power sources and a lock-on slide switch for continuous operation. The spindle lock allows quick wheel changes. It includes a side handle, wheel guard, and wrench, and weighs just 3.5 lbs, reducing fatigue during extended use. This versatile grinder is suitable for metalworking, concrete grinding, and more.