Learn More
Electron Microscopy Sciences Lapping & Polishing Fixture Angled Mounting Block, 10°
SDP
Catalog No. 50980980
Encompass
1" (25 mm) diameter stainless steel mounting blocks for lapping and polishing fixtures. For use with models 150, 151, and 155.
Angle: 10°
Use: Models 150, 151, 155
Catalog No. 50-980-980 Supplier Electron Microscopy Sciences Supplier No. 5015320
1" (25 mm) diameter stainless steel mounting blocks for lapping and polishing fixtures. For use with models 150, 151, and 155.
Angle: 10°
Use: Models 150, 151, 155