Learn More
Electron Microscopy Sciences Lapping & Polishing Fixture Angled Mounting Block, 90 °
SDP
Catalog No. 50980985
Encompass
1" (25 mm) diameter stainless steel mounting blocks for lapping and polishing fixtures. For use with models 150, 151, and 155.
Angle: 90°
Use: Models 150, 151, 155
Catalog No. 50-980-985 Supplier Electron Microscopy Sciences Supplier No. 5015325
1" (25 mm) diameter stainless steel mounting blocks for lapping and polishing fixtures. For use with models 150, 151, and 155.
Angle: 90°
Use: Models 150, 151, 155