Learn More
Electron Microscopy Sciences Angle Shield (New Style)
SDP
Catalog No. 5024537
Encompass_Preferred
Splash Protecting in Standing or Sitting Position Provides good protection personal from pipetting or pouring chemicals. An extra wide, angled viewing area and low bend allow the shield to be used from a standing or sitting position. Made from 6.4 mm (1/4") thick clear acrylic with non-skid feet for stability. Not recommended for Beta use.
Catalog No. 50-245-37 Supplier Electron Microscopy Sciences Supplier No. 64464
Splash Protecting in Standing or Sitting Position Provides good protection personal from pipetting or pouring chemicals. An extra wide, angled viewing area and low bend allow the shield to be used from a standing or sitting position. Made from 6.4 mm (1/4") thick clear acrylic with non-skid feet for stability. Not recommended for Beta use.