Learn More
Electron Microscopy Sciences EMS Flat-Tip Tweezers Style 128, 105 mm OAL
SDP
Catalog No. 5023970
Encompass_Preferred
Flat-Tip Tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. The tweezers have serrated grips. Thin and square tips, curved.
OAL: 105 mm
Catalog No. 50-239-70 Supplier Electron Microscopy Sciences Supplier No. 72972AP
Flat-Tip Tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements. The tweezers have serrated grips. Thin and square tips, curved.
OAL: 105 mm