Learn More
Fine Science Tools Cover Glass Forceps, Stainless Steel, Smooth, Angled, 10.5 cm
SDP
Catalog No. NC9923046
Encompass
Tip Shape: Angled/ Tips: Smooth/ Tip Dimensions: 4 x 0.6mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm
Catalog No. NC9923046 Supplier Fine Science Tools Supplier No. 1107310
Tip Shape: Angled/ Tips: Smooth/ Tip Dimensions: 4 x 0.6mm/ Alloy / Material: Stainless Steel/ Length: 10.5cm