Learn More
MSC Vikan Angled Dump Tub, High-density Polyethylene, 101.85 Gallon, with Drain Plug
Catalog No. 502838865
Encompass
Vikan angled dump tubs offer a combination of durability and versatility. Manufactured from high-density polyethylene, they can be used for transporting, freezing, and storing. This model features an angled edge for easy tipping and emptying, and it includes a drain plug. These tubs meet all USDA specifications and are Halal and Kosher compliant, making them suitable for various industrial applications including food processing.
  • Angled edge for easy tipping and emptying
  • Includes a drain plug
  • Constructed from high-density polyethylene for durability
  • Suitable for transporting, freezing, and storing
  • Meets all USDA specifications
  • Halal and Kosher compliant
  • Ideal for meat, seafood, poultry, commercial fishing, confectionery, bakery, and grain applications
Catalog No. 50-283-8865 Supplier MSC Supplier No. 45062593
Vikan angled dump tubs offer a combination of durability and versatility. Manufactured from high-density polyethylene, they can be used for transporting, freezing, and storing. This model features an angled edge for easy tipping and emptying, and it includes a drain plug. These tubs meet all USDA specifications and are Halal and Kosher compliant, making them suitable for various industrial applications including food processing.
  • Angled edge for easy tipping and emptying
  • Includes a drain plug
  • Constructed from high-density polyethylene for durability
  • Suitable for transporting, freezing, and storing
  • Meets all USDA specifications
  • Halal and Kosher compliant
  • Ideal for meat, seafood, poultry, commercial fishing, confectionery, bakery, and grain applications