Learn More
Fine Science Tools London Forceps, Stainless Steel, Serrated, 16 cm
SDP
Catalog No. NC9574453
Encompass
Tip Shape: Angled 45 deg./ Tips: Serrated/ Tip Dimensions: 1 x 0.9mm/ Alloy / Material: Stainless Steel/ Length: 16cm
Catalog No. NC9574453 Supplier Fine Science Tools Supplier No. 1108002
Tip Shape: Angled 45 deg./ Tips: Serrated/ Tip Dimensions: 1 x 0.9mm/ Alloy / Material: Stainless Steel/ Length: 16cm