Learn More
Bellco Glass, Inc. Angled Harvest Pipe Assembly , 15L For 18mm Port
SDP
Catalog No. 501946486
Encompass_Preferred
Stainless steal 12mm small volume harvesting pipe with 90 degree angle for sampling contents in 3L bioreactor vessel. Fits 18mm port on 3L metric headplate. Tube dimesnions: .375" OD x .305" ID x 18 1/2" with 2" bend. Inseration Length: 15 1/2". Supply O-ring, Washer, SST nut. MADE IN THE USA
Catalog No. 50-194-6486 Supplier Bellco Glass, Inc. Supplier No. 790915090
Stainless steal 12mm small volume harvesting pipe with 90 degree angle for sampling contents in 3L bioreactor vessel. Fits 18mm port on 3L metric headplate. Tube dimesnions: .375" OD x .305" ID x 18 1/2" with 2" bend. Inseration Length: 15 1/2". Supply O-ring, Washer, SST nut. MADE IN THE USA