Learn More
Bellco Glass, Inc. Angled Harvest Pipe Assembly , 3L Large Volume
SDP
Catalog No. 501946424
Encompass_Preferred
Stainless steal 12mm harvesting pipe with 90 degree angle for sampling contents in 3L bioreactor vessel. Fits 18mm port on 3L metric headplate.. MADE IN THE USA
Catalog No. 50-194-6424 Supplier Bellco Glass, Inc. Supplier No. 790900009
Stainless steal 12mm harvesting pipe with 90 degree angle for sampling contents in 3L bioreactor vessel. Fits 18mm port on 3L metric headplate.. MADE IN THE USA