Learn More
Bellco Glass, Inc. Angled Harvest Pipe Assembly , 7L Large Volume
SDP
Catalog No. 501946461
Encompass_Preferred
Stainless steal 12mm harvesting pipe with 90 degree angle for sampling contents in 3L bioreactor vessel. Fits 18mm port on 3L metric headplate. Dimensions: 12mm OD x 16 3/4" Long x 3 5/8" (angled).. MADE IN THE USA
Catalog No. 50-194-6461 Supplier Bellco Glass, Inc. Supplier No. 790900101
Stainless steal 12mm harvesting pipe with 90 degree angle for sampling contents in 3L bioreactor vessel. Fits 18mm port on 3L metric headplate. Dimensions: 12mm OD x 16 3/4" Long x 3 5/8" (angled).. MADE IN THE USA