Learn More
Electron Microscopy Sciences Lapping & Polishing Fixture Angled Mounting Block, 5 °
SDP
Catalog No. 50980979
Encompass
1" (25 mm) diameter stainless steel mounting blocks for lapping and polishing fixtures. For use with models 150, 151, and 155.
Angle: 5°
Use: Models 150, 151, 155
Catalog No. 50-980-979 Supplier Electron Microscopy Sciences Supplier No. 5015307
1" (25 mm) diameter stainless steel mounting blocks for lapping and polishing fixtures. For use with models 150, 151, and 155.
Angle: 5°
Use: Models 150, 151, 155