Learn More
Electron Microscopy Sciences Angled Tube Rack Stand, 1 each
SDP
Catalog No. 501927374
Encompass_Preferred
Easily view samples and tube contents at a 30 degree angle. Help alleviate repetitive pipetting related fatige and injuries by creating a comfortable work station. Neoprene pad holds rack in place while non-slip feet prevent stand from sliding on benchtops. Stand is compatible with most tube racks. Holds racks up to 5.50" high and 8.50" wide.
Catalog No. 50-192-7374 Supplier Electron Microscopy Sciences Supplier No. 7098701
Easily view samples and tube contents at a 30 degree angle. Help alleviate repetitive pipetting related fatige and injuries by creating a comfortable work station. Neoprene pad holds rack in place while non-slip feet prevent stand from sliding on benchtops. Stand is compatible with most tube racks. Holds racks up to 5.50" high and 8.50" wide.