Learn More
Medchemexpress LLC Angoline hydrochloride | 21080-31-9 | 5 MG
SDP
Catalog No. 5000417568
Encompass_Preferred
Angoline hydrochloride is a potent and selective IL6/STAT3 signaling pathway inhibitor with an IC50 of 11.56 μM. Angoline hydrochloride inhibits STAT3 phosphorylation and its target gene expression, and inhibits cancer cell proliferation.
  • Potent and selective IL6/STAT3 signaling pathway inhibitor
  • IC50 of 11.56 μM
  • Inhibits STAT3 phosphorylation
  • Inhibits its target gene expression
  • Inhibits cancer cell proliferation
  • Purity: >98%
Catalog No. 50-004-17568 Supplier Medchemexpress LLC Supplier No. HYN7674A1MG
Angoline hydrochloride is a potent and selective IL6/STAT3 signaling pathway inhibitor with an IC50 of 11.56 μM. Angoline hydrochloride inhibits STAT3 phosphorylation and its target gene expression, and inhibits cancer cell proliferation.
  • Potent and selective IL6/STAT3 signaling pathway inhibitor
  • IC50 of 11.56 μM
  • Inhibits STAT3 phosphorylation
  • Inhibits its target gene expression
  • Inhibits cancer cell proliferation
  • Purity: >98%