Learn More
Medchemexpress LLC HY-N0062 5mg Medchemexpress, Angoroside C CAS:115909-22-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN00625MG
Encompass_Preferred
Medchemexpress, HY-N0062 5mg Angoroside C CAS:115909-22-3 Angoroside C, a phenylpropanoid glycoside isolated from Radix Scrophulariae, has beneficial effects against ventricular remodeling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0218