Learn More
AdipoGen anti-Angiopoietin-2 (human), mAb (rec.) (blocking) (Angy-1-4) (preservative free) (100 µg)
SDP
Supplier:  AdipoGen AG27B0015PFC100
Encompass_Preferred
Recombinant Antibody. Recognizes human angiopoietin-2. Does not recognize mouse angiopoietin-2 and human angiopoietin-1. Species cross-reactivity: Human. Clone: Angy-1-4. Isotype: Human IgG2lambda. Applications: ELISA, FUNC (Blocking). Host: CHO cells. Liquid. In PBS containing 10% glycerol. Ang-1 activates Tie-2 signaling on endothelial cells to promote chemotaxis, cell survival, cell sprouting, vessel growth and stabilization. Ang-2 has been identified as a secreted protein ligand of Tie-2 and has alternatively been reported to be an antagonist for Ang-1 induced Tie-2 signaling as well as an agonist for Tie-2 signaling, depending on the cell context. anti-Angiopoietin-2 (human), mAb (rec.) (blocking) (Angy-1-4) (preservative free) is an antibody developed by antibody phage display technology using a human naive antibody gene library.
Catalog No. 50-205-3036