Learn More
Aviva Systems Biology Corp ANGPT1 Antibody - N-terminal region (ARP63106_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63106_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANGPT1. It was validated on Western Blot. . Alternate names: AGP1, AGPT, ANG1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-879