Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ANGPT2 (Human) ELISA Kit

Catalog No. p-4392803 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Plasma, Serum, Tissue Sample, Urine
CCS, Plasma, Serum, Urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-728 CCS, Plasma, Serum, Tissue Sample, Urine
89-028-578 CCS, Plasma, Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-728 Supplier Abnova™ Supplier No. KA3021
Only null left
Add to Cart
Add to Cart

In-Vitro Enzyme-Linked Iimmunosorbent assay for the quantitative measurement of Human ANGPT2

Specifications

Calibration Range 8 to 150ng/mL
Conjugate HRP
Cross Reactivity Human
Description ANGPT2 (Human) ELISA Kit is intended for the quantitative measurement of human ANGPT2.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes One 96-well plate pre-coated with anti-human ANGII antibody, Standard (180pg/mL) (0.5mL), Standard diluent buffer (1.5mL), Wash buffer (30x), Dilution: 1:30 (20mL), Sample diluent buffer (6mL), HRP conjugated anti-human ANGII antibody (RTU) (6mL), Stop solution (6mL), TMB substrate A (6mL), TMB substrate B (6mL), Plate sealer (2 slices), Hermetic bag (1 bag)
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma, Serum, Tissue Sample, Urine
Sample Volume 10 μL
Content And Storage Store at 4°C for 6 months.
For Use With (Application) Quantification
Product Type ANGPT2 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.