Learn More
Aviva Systems Biology Corp ANGPTL1 Antibody - N-terminal region (ARP64602_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64602_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANGPTL1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human ANGPTL1. Alternate names: ANG3, ANGPT3, ARP1, AngY, KIAA0351, UNQ162, dJ595C2.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-311