Learn More
IBL America ANGPTL2
Supplier:  IBL America 27745
Encompass_Preferred
Solid phase sandwich ELISA for the determination of human ANGPTL2 in serum, EDTA-plasma and cell culture supernatant. For research use only, not for use in diagnostic procedures.Tetra Methyl Benzidine (TMB) is used as coloring agent (Chromogen). The strength of coloring is in proportion to the quantities of human ANGPTL2.
Catalog No. 50-136-6543