Learn More
AdipoGen ANGPTL2 human pAb
SDP
Supplier:  AdipoGen AG25A0068C100
Encompass_Preferred
Polyclonal Antibody. Recognizes the fibrinogen-like domain of human ANGPTL2. Detects a band of ~57kDa by Western blot. Cross-reacts with full-length human ANGPTL2. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. ANGPTL2 stimulates expansion of HSCs. ANGPTL2 plays an important role in the pathogenesis of diabetic glomerulopathy. The upregulation of ANGPTL2 in diabetic glomerulopathy shows a close relationship to abnormal microvasculature and endothelial inflammation.
Catalog No. 50-256-3245