Learn More
Aviva Systems Biology Corp ANGPTL3 Antibody - N-terminal region (ARP42411_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42411_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANGPTL3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ANGPTL3. Alternate names: ANGPT5, FHBL2ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-001