Learn More
AdipoGen ANGPTL3 CCD h pAb
SDP
Supplier:  AdipoGen AG25A0060C100
Encompass_Preferred
Polyclonal Antibody. Recognizes the coiled-coil domain of human ANGPTL3. Detects a band of ~29kDa by Western blot. Weakly cross-reacts with human full lenght ANGPTL3. Source Rabbit. Applications ELISA, IHC, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. ANGPTL3 regulates angiogenesis and also directly regulates lipid, glucose, and energy metabolism.
Catalog No. 50-256-3252