Learn More
AdipoGen ANGPTL3 human ELISA Kit
SDP
Supplier:  AdipoGen AG45A0014YEKKI01
Encompass_Preferred
ELISA Assay. Detects human ANGPTL3. Does not cross-react with mouse ANGPTL3, human ANGPTL4, human ANGPTL6, human ANGPTL7 or human ANGPTL2. Colorimetric assay. Sample Types Cell Culture Supernatant, Plasma, Serum. Range 0.156 to 10ng/ml. Sensitivity 75pg/ml. Seven proteins containing an N-terminal coiled-coil domain and a C-terminal fibrinogen-like domain, both characteristics of angiopoietins, have been identified and therefore designated angiopoietin-like proteins 1-7 ANGPTL 1-7. ANGPTL3 and ANGPTL6 Angiopoietin-related growth factor AGF, among others, function to regulate angiogenesis and also appear to directly regulate lipid, glucose, and energy metabolism. ANGPTL3 plasma levels are increased in a mouse model of diabetes mellitus.
Catalog No. 50-256-2742