Learn More
AdipoGen ANGPTL3 mouse pAb
SDP
Supplier:  AdipoGen AG25A0070C100
Encompass_Preferred
Polyclonal Antibody. Recognizes mouse ANGPTL3. Detects a band of ~64kDa by Western blot predicted ~54kDa. Weakly cross-reacts with human ANGPTL3. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. ANGPTL3 regulates angiogenesis and also directly regulates lipid, glucose, and energy metabolism.
Catalog No. 50-256-3243