Learn More
AdipoGen ANGPTL3 mouse rec
SDP
Catalog No. 502563006
Encompass_Preferred
Protein. Mouse ANGPTL3 aa 17-455 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. ANGPTL3 Angiopoietin-like protein 3 regulates angiogenesis and also directly regulates lipid, glucose, and energy metabolism.
Catalog No. 50-256-3006 Supplier AdipoGen Supplier No. AG40A0082C010
Protein. Mouse ANGPTL3 aa 17-455 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. ANGPTL3 Angiopoietin-like protein 3 regulates angiogenesis and also directly regulates lipid, glucose, and energy metabolism.