Learn More
AdipoGen ANGPTL3h mAb Kairos-37
SDP
Supplier:  AdipoGen AG20A0039C050
Encompass_Preferred
Monoclonal Antibody. Recognizes the fibrinogen-like domain FLD of human ANGPTL3. Detects a band of ~68kDa and a cleaved band of ~35kDa by Western blot. Does not cross-react with other ANGPTL family proteins. Isotype Mouse IgG1kappa. Clone Kairos-37. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. ANGPTL3 regulates angiogenesis and also directly regulates lipid, glucose, and energy metabolism.
Catalog No. 50-256-3390