Learn More
AdipoGen anti-ANGPTL4 (coiled-coil domain) (human), mAb (Kairos4-153AD) (50 µg)
SDP
Supplier:  AdipoGen AG20A0046C050
Encompass_Preferred
Monoclonal Antibody. Recognizes the coiled-coil domain (CCD) of human ANGPTL4. Detects a band of ~19kDa by Western blot. Isotype: Mouse IgG1kappa. Clone: Kairos4-153AD. Applications: ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. ANGPTL4 mainly expressed in endothelial cells (hypoxia-induced). Regulates angiogenesis and modulates tumorgenesis and directly regulates lipid, glucose, and energy metabolism. Inhibits proliferation, migration, and tubule formation of endothelial cells and reduces vascular leakage. ANGPTL4 is a protein consisting of an N-terminal coiled-coil domain and a C-terminal fibrinogen-like domain (FLD). Both domains have distinct biological functions. The coiled-coil domain is responsible for the inhibitory effects on lipoprotein lipase (LPL) converting the active form of LPL into an inactive form, and the FLD domain mediates its antiangiogenic functions.
Catalog No. 50-205-2844