Learn More
AdipoGen ANGPTL4 FLD h rec
SDP
Catalog No. 502563022
Encompass_Preferred
Protein. Fibrinogen-like domain of human ANGPTL4 aa 166-406 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. ANGPTL4 Angiopoietin-like protein 4 mainly expressed in endothelial cells hypoxia-induced. Regulates angiogenesis and modulates tumorigenesis and directly regulates lipid, glucose, and energy metabolism. Inhibits proliferation, migration, and tubule formation of endothelial cells and reduces vascular leakage. ANGPTL4 is a protein consisting of an N-terminal coiled-coil domain and a C-terminal fibrinogen-like domain FLD. Both domains have distinct biological functions. The coiled-coil domain is responsible for the inhibitory effects on lipoprotein lipase LPL converting the active form of LPL into an inactive form, and the FLD domain mediates its antiangiogenic functions.
Catalog No. 50-256-3022 Supplier AdipoGen Supplier No. AG40A0070C010
Protein. Fibrinogen-like domain of human ANGPTL4 aa 166-406 is fused at the N-terminus to a FLAG -tag. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. ANGPTL4 Angiopoietin-like protein 4 mainly expressed in endothelial cells hypoxia-induced. Regulates angiogenesis and modulates tumorigenesis and directly regulates lipid, glucose, and energy metabolism. Inhibits proliferation, migration, and tubule formation of endothelial cells and reduces vascular leakage. ANGPTL4 is a protein consisting of an N-terminal coiled-coil domain and a C-terminal fibrinogen-like domain FLD. Both domains have distinct biological functions. The coiled-coil domain is responsible for the inhibitory effects on lipoprotein lipase LPL converting the active form of LPL into an inactive form, and the FLD domain mediates its antiangiogenic functions.