Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ANGPTL4 ELISA Kit

Catalog No. EHANGPTL4X5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHANGPTL4X5 5 x 96 Tests
EHANGPTL4 96 Tests
50-274-1175 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EHANGPTL4X5 Supplier Invitrogen™ Supplier No. EHANGPTL4X5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

The Human Angiopoietin Like4 (Hu ANGPTL4) ELISA quantitates Hu ANGPTL4 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ANGPTL4. Principle of the method The Human ANGPTL4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Angiopoietin-like protein 4 (ANGPTL4), is a secreted protein selectively expressed in adipose tissue, liver, and placenta1-3 that plays a variety of roles in vivo, ranging from adipogenesis to angiogenesis to carcinogenesis. The expression of ANGPTL4 is also under nutritional and hormonal control. ANGPTL4 has been observed to induce a strong pro-angiogenic response independent of vascular endothelial growth factor (VEGF), and its expression has been described in hypoxic human tissues as well as a variety of cancers, including liposarcoma, hepatocellular carcinoma, and conventional renal cell carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BY76
Assay Range 20-20000 pg/mL
Assay Sensitivity 20 pg/mL
Calibration Range 20 pg/mL to 2,0000 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF- 4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF
Sufficient For 5 x 96 Assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 425O18-1; angiopoietin like 4; angiopoietin-like 4; angiopoietin-like protein 4; Angiopoietin-related protein 4; ANGPTL2; ANGPTL4; ANGPTL4 C-terminal chain; ANGPTL4 N-terminal chain; ARP4; Bk89; Farp; Fasting-i; Fasting-induced adipose factor; FIAF; fibrinogen/angiopoietin-related protein; HARP; hepatic angiopoietin-related protein; hepatic fibrinogen/angiopoietin-related protein; HFARP; major histocompatibility complex region NG27; Ng27; NL2; peroxisome proliferator-activated receptor (PPAR) gamma induced angiopoietin-related protein; PGAR; Pgarg; Pp1158; PPARG angiopoietin related protein; PSEC0166; Secreted protein Bk89; TGQTL; UNQ171; UNQ171/PRO197425O18-1; angiopoietin like 4; angiopoietin-like 4; angiopoietin-like protein 4; Angiopoietin-related protein 4; ANGPTL2; ANGPTL4; ANGPTL4 C-terminal chain; ANGPTL4 N-terminal chain; ARP4; Bk89; Farp; Fasting-i; Fasting-induced adipose factor; FIAF; fibrinogen/angiopoietin-related protein; HARP; hepatic angiopoietin-related protein; hepatic fibrinogen/angiopoietin-related protein; HFARP; major histocompatibility complex region NG27; Ng27; NL2; peroxisome proliferator-activated receptor (PPAR) gamma induced angiopoietin-related protein; PGAR; Pgarg; Pp1158; PPARG angiopoietin related protein; PSEC0166; Secreted protein Bk89; TGQTL; UNQ171; UNQ171/PRO197; ARP4; FIAF; HARP; HFARP; NL2; PGAR; TGQTL; UNQ171; pp1158
Gene ID (Entrez) 51129
Gene Symbol ANGPTL4
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Angiopoietin-like protein 4, Hepatic fibrinogen/angiopoietin-related protein, HFARP
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Angiogenesis
Sample Volume Plasma 33 μL, Serum 33 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym PP1158, ANGPTL2, ARP4, FIAF, HARP, HFARP, NL2, PGAR, TGQTL, UNQ171, pp1158
Target ANGPTL4
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.