Learn More
AdipoGen ANGPTL5 CCD h pAb
SDP
Supplier:  AdipoGen AG25A0069C100
Encompass_Preferred
Polyclonal Antibody. Recognizes the coiled-coil domain of human ANGPTL5. Detects a band of ~14kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. ANGPTL5 is mainly expressed in adult human heart. It regulates blood vessel regeneration, angiogenesis and most probably modulates tumorgenesis.
Catalog No. 50-256-3244