Learn More
AdipoGen ANGPTL6 human pAb
SDP
Supplier:  AdipoGen AG25A0030C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human ANGPTL6. Detects a band of ~52kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. ANGPTL6 regulates angiogenesis and also directly regulates lipid, glucose, and energy metabolism. The serum levels of ANGPTL6 are significantly increased in obese humans, patients with type 2 diabetes mellitus T2DM, and in metabolic syndrome.
Catalog No. 50-256-3282