Learn More
Aviva Systems Biology Corp ANGPTL7 Antibody - middle region (ARP49626_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49626_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANGPTL7. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ANGPTL7. Alternate names: AngX, CDT6, RP4-647M16.2, dJ647M16.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-439