Learn More
AdipoGen anti-ANGPTL7 (human), mAb (Kairos 397-7) (100 µg)
SDP
Supplier:  AdipoGen AG20A0055C100
Encompass_Preferred
Monoclonal Antibody. Recognizes human ANGPTL7. Detects a band of ~45kDa by Western blot. Weakly cross-reacts with the fibrinogen-like domain (FLD) of human ANGPTL7. Isotype: Mouse IgG1kappa. Clone: Kairos 397-7. Applications: ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. ANGPTL7 is expressed in various tissues and regulates angiogenesis. Human ANGPTL7 was characterized as a potent target gene of the WNT/ beta-catenin signaling pathway, and is a pharmacogenomics target in the fields of oncology and regenerative medicine. Overexpression of ANGPTL7 increases collagen expression and might exert a pathogenic role in glaucoma.
Catalog No. 50-205-2853