Learn More
ACROBiosystems Recombinant Protein;Human ANGPTL7 / CDT6 Protein, Mouse IgG2a Fc Tag;HEK293;1MG;AN7,ANGPTL7,CDT6
Catalog No. 501658060
Encompass_Preferred
Recombinant Protein;Human ANGPTL7 / CDT6 Protein, Mouse IgG2a Fc Tag;1MG;Human ANGPTL7, Mouse IgG2a Fc Tag (AN7-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Pro 346 (Accession # O43827-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 64.1 kDa. The protein migrates as 60 kDa and 76 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LILRB2, Fc Tag (Cat. No. CDD-H5259) at 1 μg/mL (100 μL/well) can bind Human ANGPTL7, Mouse IgG2a Fc Tag (Cat. No. AN7-H5252) with a linear range of 0.313-1.25 μg/mL (Routinely tested).;Immobilized Human LILRB4, Fc Tag (Cat. No. LI4-H5259) at 1 μg/mL (100 μL/well) can bind Human ANGPTL7, Mouse IgG2a Fc Tag (Cat. No. AN7-H5252) with a linear range of 156-625 ng/mL (Routinely tested).
Catalog No. 50-165-8060 Supplier ACROBiosystems Supplier No. AN7H52521MG
Recombinant Protein;Human ANGPTL7 / CDT6 Protein, Mouse IgG2a Fc Tag;1MG;Human ANGPTL7, Mouse IgG2a Fc Tag (AN7-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Pro 346 (Accession # O43827-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 64.1 kDa. The protein migrates as 60 kDa and 76 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LILRB2, Fc Tag (Cat. No. CDD-H5259) at 1 μg/mL (100 μL/well) can bind Human ANGPTL7, Mouse IgG2a Fc Tag (Cat. No. AN7-H5252) with a linear range of 0.313-1.25 μg/mL (Routinely tested).;Immobilized Human LILRB4, Fc Tag (Cat. No. LI4-H5259) at 1 μg/mL (100 μL/well) can bind Human ANGPTL7, Mouse IgG2a Fc Tag (Cat. No. AN7-H5252) with a linear range of 156-625 ng/mL (Routinely tested).