Learn More
AdipoGen ANGPTL8 msFc hrec
SDP
Catalog No. 502562912
Encompass_Preferred
Protein. Mouse ANGPTL8 Betatrophin 22-198 is fused at the C-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS 0.02% CHAPS. Purity >90% SDS-PAGE. ANGPTL8 Angiopoietin-like protein 8 RIFL Lipasin Betatrophin is a newly discovered secreted protein of 198 aa that was proposed to promote beta cell proliferation and improve glucose tolerance in mice. ANGPTL8 may also function in inhibition of lipase activity and on serum triglyceride regulation. ANGPTL8 is expressed in the liver and in white and brown adipose tissue of mice. In humans, ANGPTL8 is found to be predominantly expressed in the liver. ANGPTL8 levels are reduced by fasting and are elevated upon insulin resistance and during pregnancy. ANGPTL8, according to preliminary data could bind to the macrophage receptor Marco and also to RTN4R, a neuronal receptor.
Catalog No. 50-256-2912 Supplier AdipoGen Supplier No. AG40B01423010
Protein. Mouse ANGPTL8 Betatrophin 22-198 is fused at the C-terminus to the Fc portion of human IgG1. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS 0.02% CHAPS. Purity >90% SDS-PAGE. ANGPTL8 Angiopoietin-like protein 8 RIFL Lipasin Betatrophin is a newly discovered secreted protein of 198 aa that was proposed to promote beta cell proliferation and improve glucose tolerance in mice. ANGPTL8 may also function in inhibition of lipase activity and on serum triglyceride regulation. ANGPTL8 is expressed in the liver and in white and brown adipose tissue of mice. In humans, ANGPTL8 is found to be predominantly expressed in the liver. ANGPTL8 levels are reduced by fasting and are elevated upon insulin resistance and during pregnancy. ANGPTL8, according to preliminary data could bind to the macrophage receptor Marco and also to RTN4R, a neuronal receptor.