Learn More
Medchemexpress LLC HY-P2230 5mg Medchemexpress, Angstrom6 CAS:220334-14-5 Purity:>98%
SDP
Catalog No. 502083081
Encompass_Preferred
Medchemexpress, HY-P2230 5mg Angstrom6 CAS:220334-14-5 Angstrom6 (A6 Peptide) is an 8 amino-acid peptide derived from single-chain urokinase plasminogen activator (scuPA) and interferes with the uPA/uPAR cascade and abrogates downstream effects. Angstrom6 binds to CD44 resulting in the inhibition of migration, invasion, and metastasis of tumor cells, and the modulation of CD44-mediated cell signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3081 Supplier Medchemexpress LLC Supplier No. HYP22305MG
Add to Cart
Add to Cart
Medchemexpress, HY-P2230 5mg Angstrom6 CAS:220334-14-5 Angstrom6 (A6 Peptide) is an 8 amino-acid peptide derived from single-chain urokinase plasminogen activator (scuPA) and interferes with the uPA/uPAR cascade and abrogates downstream effects. Angstrom6 binds to CD44 resulting in the inhibition of migration, invasion, and metastasis of tumor cells, and the modulation of CD44-mediated cell signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.