Learn More
Selleck Chemical LLC Anhydroicaritin S9080-1MG
SDP
Catalog No. 5000025231
Encompass_Preferred
Anhydroicaritin (Cycloicaritin) a prenylated flavonoid natural product is commonly recognized as one of the effective compounds of Epimedii Herba and exhibits a variety of biological activities such as activation of cancer cell apoptosis and inhibition of growth protection against beta amyloid-induced neurotoxicity and promotion of neuronal and cardiac cellular differentiation It is a potent inhibitor of transcription factor SREBPs
Catalog No. 50-000-25231 Supplier Selleck Chemical LLC Supplier No. S90801MG
Anhydroicaritin (Cycloicaritin) a prenylated flavonoid natural product is commonly recognized as one of the effective compounds of Epimedii Herba and exhibits a variety of biological activities such as activation of cancer cell apoptosis and inhibition of growth protection against beta amyloid-induced neurotoxicity and promotion of neuronal and cardiac cellular differentiation It is a potent inhibitor of transcription factor SREBPs