Learn More
Cayman Chemical Anhydrotetracycline (hydrochloride), 13803-65-1, 100 mg
SDP
Supplier:  Cayman Chemical 10009542100
Encompass_Preferred
Molecular Formula C22H22N2O7 • HCl, Purity ≥98%, Formula Weight 462.9, Synonyms: 4-(dimethylamino)-1,4S,4aS,5,12,12aS-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-monohydrochloride-2-naphthacenecarboxamide; A powerful effector in both the TetR and revTetR transcriptional regulator systems, binding the Tet repressor 35-fold more strongly than Tet; does not act a general inhibitor of translation and is a poor antibiotic.
Catalog No. NC0526524