Learn More
Takara Bio Anhydrotetracycline, 200 ul
SDP
Catalog No. NC9451757
Encompass
Anhydrotetracycline is an antibiotic derivative of tetracycline. It is effective at lower concentrations than either tetracycline or doxycycline and is also less toxic to E. coli. It is thus a suitable antibiotic for use with the PROTet Vectors or other vectors or systems designed upon elements of the (Tn10) tet operon. 200ul
Catalog No. NC9451757 Supplier Takara Bio Supplier No. 631310
Anhydrotetracycline is an antibiotic derivative of tetracycline. It is effective at lower concentrations than either tetracycline or doxycycline and is also less toxic to E. coli. It is thus a suitable antibiotic for use with the PROTet Vectors or other vectors or systems designed upon elements of the (Tn10) tet operon. 200ul