Learn More
IBA LifeSciences Anhydrotetracycline; 25 mg
Supplier:  IBA LifeSciences 20401002
Encompass_Preferred
Anhydrotetracycline is a derivative of tetracycline and an efficient inducer of the tetracycline promoter present in E. coli expression vectors pASK-IBA and pASG-IBA. It does not exhibit antibiotic activity within our recommended concentrations.
Catalog No. 50-210-0126