Learn More
Apexbio Technology LLC Anhydrotetracycline (hydrochloride) 13803-65-1 250mg
SDP
Catalog No. 5000064078
Encompass_Preferred
Anhydrotetracycline hydrochloride (ATC) is an effector molecule utilized in tetracycline-controlled gene expression systems modulating the DNA-binding activity of tetracycline repressor protein (TetR) and its engineered variant reverse TetR (revTetR) TetR binds to operator DNA sequences inhibiting transcription ATC binding induces TetR conformational changes dissociating it from operator DNA enabling gene expression Conversely ATC acts as a corepressor in revTetR systems enhancing its DNA-binding inhibition ATC displays approximately 35-fold greater affinity towards TetR compared to tetracycline with an IC50 near 0 4 M employed in inducible gene expression studies in biological research contexts
Catalog No. 50-000-64078 Supplier Apexbio Technology LLC Supplier No. C4291250
Add to Cart
Add to Cart
Anhydrotetracycline hydrochloride (ATC) is an effector molecule utilized in tetracycline-controlled gene expression systems modulating the DNA-binding activity of tetracycline repressor protein (TetR) and its engineered variant reverse TetR (revTetR) TetR binds to operator DNA sequences inhibiting transcription ATC binding induces TetR conformational changes dissociating it from operator DNA enabling gene expression Conversely ATC acts as a corepressor in revTetR systems enhancing its DNA-binding inhibition ATC displays approximately 35-fold greater affinity towards TetR compared to tetracycline with an IC50 near 0 4 M employed in inducible gene expression studies in biological research contexts