Learn More
AdipoGen Anhydrotetracycline hydrochloride, 13803-65-1, MFCD00151453, 500 mg
SDP
Supplier:  AdipoGen CDXA0197M500
Encompass_Preferred
MF: C22H22N2O7 . HCl, MW: 462.88, ≥98%, Appearance: Yellow powder. A tetracycline derivative exhibiting no antibiotic activity. A useful effector of tetracycline-dependent gene expression in bacterial expression systems.
Catalog No. 50-595-757